N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-16, SB-16)

Lieferant: Molekula

Synonyme: Palmityldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, Hexadecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(palmityl)ammonio]propane-1-sulphonate, Palmityl sulphobetaine, 3-(Hexadecyldimethylammonio)propane-1-sulphonate

57354850-25G
MOLE57354850-25GEA 140 EUR
MOLE57354850-25G
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-16, SB-16)
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-16, SB-16)
Formel: C₂₁H₄₅NO₃S
Molecular Weight: 391,6500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036911
CAS-Nummer: 2281-11-0
EINECS: 218-918-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR