trans-2,5-Dimethylpiperazin

Lieferant: Molekula

57459985-25G
MOLE57459985-25GEA 175 EUR
MOLE57459985-25G
trans-2,5-Dimethylpiperazin
trans-2,5-Dimethylpiperazin
Formel: C₆H₁₄N₂
Molecular Weight: 114,19 g/mol
MDL: MFCD00070516
CAS-Nummer: 2815-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR