(R)-(+)-4-Isopropyl-2-oxazolidinon

Lieferant: Molekula

Synonyme: (R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone

57944281-1G
MOLE57944281-1GEA 0 EUR
MOLE57944281-1G
(R)-(+)-4-Isopropyl-2-oxazolidinon
(R)-(+)-4-Isopropyl-2-oxazolidinon
Formel: C₆H₁₁NO₂
Molecular Weight: 129,16 g/mol
MDL: MFCD00075172
CAS-Nummer: 95530-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR