2,4-Dimethoxypyrimidin

Lieferant: Molekula

58183615-1G
MOLE58183615-1GEA 0 EUR
MOLE58183615-1G
2,4-Dimethoxypyrimidin
2,4-Dimethoxypyrimidine
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
Siedepunkt: 205 °C (1013 hPa)
Schmelzpunkt: 17,5 °C
MDL: MFCD00056028
CAS-Nummer: 3551-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR