Hydrindantin Dihydrat
Lieferant: Molekula
Synonyme:
(2,2'-Biindan)-1,1',3,3'-tetrone, 2,2'-dihydroxy-, 1,1'-dihydrate (7CI), 2,2',3,3,3',3'-Hexahydroxy-2,2'-biindan-1,1'-dione
58435547
MOLE58435547EA
0
EUR
MOLE58435547
Hydrindantin Dihydrat
Hydrindantin Dihydrat
Formel:
C₁₈H₁₀O₆·2H₂O Molecular Weight: 358,30 g/mol |
MDL:
MFCD00149242 CAS-Nummer: 5950-69-6 EINECS: 227-713-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...