5-(3-Pyridyl)-1H-tetrazol

Lieferant: Molekula

Synonyme: 3-(2H-Tetrazol-5-yl)pyridine

58528284-1G
MOLE58528284-1GEA 0 EUR
MOLE58528284-1G
5-(3-Pyridyl)-1H-tetrazol
5-(3-Pyridyl)-1H-tetrazole
Formel: C₆H₅N₅
Molecular Weight: 147,14 g/mol
MDL: MFCD00040281
CAS-Nummer: 3250-74-6
EINECS: 426-810-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR