4-Nitro-m-kresol
Lieferant: Molekula
|
Achtung
|
Synonyme:
3-Methyl-4-nitrophenol, 1-Hydroxy-3-methyl-4-nitrobenzol, 5-Hydroxy-2-nitrotoluol
58558107-100G
MOLE58558107-100GEA
0
EUR
MOLE58558107-100G
4-Nitro-m-kresol
4-Nitro-m-kresol
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Schmelzpunkt: 126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007333 CAS-Nummer: 2581-34-2 EINECS: 219-952-5 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...