3-(Hydroxymethyl)phenylboronsäure

Lieferant: Molekula

Synonyme: 3-(Hydroxymethyl)benzeneboronic acid, (3-Hydroxymethyl)phenylboronic acid

59697211-1G
MOLE59697211-1GEA 0 EUR
MOLE59697211-1G
3-(Hydroxymethyl)phenylboronsäure
3-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
MDL: MFCD01317846
CAS-Nummer: 87199-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR