3-Cyanbenzoesäure
Lieferant: Molekula
|
Achtung
|
60148293-25G
MOLE60148293-25GEA
0
EUR
MOLE60148293-25G
3-Cyanbenzoesäure
3-Cyanbenzoesäure
Formel:
C₈H₅NO₂ Molecular Weight: 147.13 g/mol Schmelzpunkt: 219 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002486 CAS-Nummer: 1877-72-1 EINECS: 217-511-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...