2'-Deoxyuridin

Lieferant: Molekula

61613969-100G 61613969-5G
MOLE61613969-100GEA 0 EUR
MOLE61613969-100G MOLE61613969-5G
2'-Deoxyuridin
2'-Deoxyuridin
Formel: C₉H₁₂N₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00006527
CAS-Nummer: 951-78-0
EINECS: 213-455-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR