(3-Chlorpropyl)dimethylammoniumchlorid

Lieferant: Molekula

61642496-500G
MOLE61642496-500GEA 0 EUR
MOLE61642496-500G
(3-Chlorpropyl)dimethylammoniumchlorid
3-Chloropropyldimethylammonium chloride
Formel: C₅H₁₃Cl₂N
MDL: MFCD00012521
CAS-Nummer: 5407-04-5
EINECS: 226-467-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR