3-Phenylpropionitril

Lieferant: Molekula

Synonyme: Hydrocinnamonitrile

61970504-25G
MOLE61970504-25GEA 0 EUR
MOLE61970504-25G
3-Phenylpropionitril
3-Phenylpropionitril
Formel: C₉H₉N
Molecular Weight: 131,18 g/mol
MDL: MFCD00001961
CAS-Nummer: 645-59-0
EINECS: 211-447-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR