1,3-Cyclopentandion

Lieferant: Molekula

Synonyme: Cyclopentane-1,3-dione

62413562-5G
MOLE62413562-5GEA 0 EUR
MOLE62413562-5G
1,3-Cyclopentandion
1,3-Cyclopentandion
Formel: C₅H₆O₂
Molecular Weight: 98,1 g/mol
Schmelzpunkt: 149…152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00001405
CAS-Nummer: 3859-41-4
EINECS: 223-372-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR