(1R,2S)-(-)-2-Amino-1,2-diphenylethanol

Lieferant: Molekula

62517904-5G
MOLE62517904-5GEA 0 EUR
MOLE62517904-5G
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol
Formel: C₁₄H₁₅NO
Molecular Weight: 213,28 g/mol
MDL: MFCD00074960
CAS-Nummer: 23190-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR