4-Nitrobenzylchlorformiat

Lieferant: Molekula

Synonyme: 4-Nitrobenzyl carbonochloridate

62613813-25G
MOLE62613813-25GEA 0 EUR
MOLE62613813-25G
4-Nitrobenzylchlorformiat
4-Nitrobenzylchlorformiat
Formel: C₈H₆ClNO₄
Molecular Weight: 215,59 g/mol
MDL: MFCD00007375
CAS-Nummer: 4457-32-3
EINECS: 224-708-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR