Indometacin

Lieferant: Molekula

62725591-25G
MOLE62725591-25GEA 116 EUR
MOLE62725591-25G
Indometacin
Indomethacin
Formel: C₁₉H₁₆ClNO₄
Molecular Weight: 357.80 g/mol
MDL: MFCD00057095
CAS-Nummer: 53-86-1
EINECS: 200-186-5
UN: 2811
ADR: 6.1 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR