2,6-Dichlorisonicotinsäure

Lieferant: Molekula

Synonyme: 2,6-Dichloropyridine-4-carboxylic acid

63506294-1G
MOLE63506294-1GEA 0 EUR
MOLE63506294-1G
2,6-Dichlorisonicotinsäure
2,6-Dichlorisonicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
MDL: MFCD00234147
CAS-Nummer: 5398-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR