Tetrabutylammoniumbromid
Lieferant: Molekula
64093691-500G
MOLE64093691-500GEA
146
EUR
MOLE64093691-500G
Tetrabutylammoniumbromid
Tetrabutylammoniumbromid
Formel:
C₁₆H₃₆BrN Molecular Weight: 322,37 g/mol Schmelzpunkt: 100…103 °C Dichte: 1,148 g/cm³ (20 °C) |
MDL:
MFCD00011633 CAS-Nummer: 1643-19-2 EINECS: 216-699-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...