(Brommethyl)cyclopropan
Lieferant: Molekula
64223627-5G
MOLE64223627-5GEA
0
EUR
MOLE64223627-5G
(Brommethyl)cyclopropan
(Brommethyl)cyclopropan
Formel:
C₄H₇Br Molecular Weight: 135 g/mol Siedepunkt: 105 °C (1013 hPa) Dichte: 1,41 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001306 CAS-Nummer: 7051-34-5 EINECS: 230-331-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...