3-Pyridinboronsäure

Lieferant: Molekula

Synonyme: 3-Pyridinylboronsäure, Pyridin-3-boronsäure

64386247-1G
MOLE64386247-1GEA 0 EUR
MOLE64386247-1G
3-Pyridinboronsäure
3-Pyridineboronic acid
Formel: C₅H₆BNO₂
Molecular Weight: 122.92 g/mol
MDL: MFCD00674177
CAS-Nummer: 1692-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR