L(-)-Methioninol

Lieferant: Molekula

Synonyme: (S)-2-Amino-4-(methylthio)butan-1-ol, L-Methioninol

64837537-1G
MOLE64837537-1GEA 0 EUR
MOLE64837537-1G
L(-)-Methioninol
L(-)-Methioninol
Formel: C₅H₁₃NOS
Molecular Weight: 135,2 g/mol
MDL: MFCD00004735
CAS-Nummer: 2899-37-8
EINECS: 220-788-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR