Bacitracin

Lieferant: Molekula

64862649-25G
MOLE64862649-25GEA 242 EUR
MOLE64862649-25G
Bacitracin
Bacitracin
Formel: C₆₆H₁₀₃N₁₇O₁₆S₁
Molecular Weight: 1421,60 g/mol
MDL: MFCD00062640
CAS-Nummer: 1405-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR