3-Aminophenylboronsäure Monohydrat
Lieferant: Molekula
Synonyme:
Dihydroxy(3-aminophenyl)boran Monohydrat, (3-Aminophenyl)metaborsäure Monohydrat, 3-Aminobenzolboronsäure Monohydrat
64941781-1G
MOLE64941781-1GEA
71.6
EUR
MOLE64941781-1G
3-Aminophenylboronsäure Monohydrat
3-Aminophenylboronsäure Monohydrat
Formel:
C₆H₈BNO₂·1H₂O Molecular Weight: 154,96 g/mol |
MDL:
MFCD00149554 CAS-Nummer: 206658-89-1 EINECS: 250-189-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...