4-Brompyridiniumchlorid
Lieferant: Molekula
|
Achtung
|
Synonyme:
4-Brompyridin Hydrochlorid, 4-Brompyridin HCl
65801625-5G
MOLE65801625-5GEA
71.6
EUR
MOLE65801625-5G
4-Brompyridiniumchlorid
4-Bromopyridinium chloride
Formel:
C₅H₅BrClN Molecular Weight: 194,46 g/mol Schmelzpunkt: 235…238 °C |
MDL:
MFCD00012828 CAS-Nummer: 19524-06-2 EINECS: 243-128-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...