4-Bromindol

Lieferant: Molekula

Synonyme: 4-Bromo-1H-indole

65921824-1G
MOLE65921824-1GEA 71.6 EUR
MOLE65921824-1G
4-Bromindol
4-Bromindol
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
MDL: MFCD00671502
CAS-Nummer: 52488-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR