6-Azaindol

Lieferant: Molekula

66418762-1G
MOLE66418762-1GEA 0 EUR
MOLE66418762-1G
6-Azaindol
6-Azaindol
Formel: C₇H₆N₂
MDL: MFCD01686899
CAS-Nummer: 271-29-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR