1-Naphthylphosphat

Lieferant: Molekula

66442790-5G
MOLE66442790-5GEA 0 EUR
MOLE66442790-5G
1-Naphthylphosphat
1-Naphthylphosphat
Formel: C₁₀H₉O₄P
CAS-Nummer: 1136-89-6
EINECS: 214-502-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR