5-Brom-2-thiophensulfonylchlorid
Lieferant: Molekula
Synonyme:
5-Brom-2-thiophensulfonsäurechlorid, 5-Bromthiophen-2-sulfonylchlorid
66601772-5G
MOLE66601772-5GEA
0
EUR
MOLE66601772-5G
5-Brom-2-thiophensulfonylchlorid
5-Brom-2-thiophensulfonylchlorid
Formel:
C₄H₂BrClO₂S₂ Molecular Weight: 261,55 g/mol |
MDL:
MFCD00084909 CAS-Nummer: 55854-46-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...