4,6-Dimethoxysalicylaldehyd
Lieferant: Molekula
Synonyme:
2-Hydroxy-4,6-dimethoxybenzaldehyde, 4,6-Dimethoxy-2-hydroxybenzaldehyde
67209584-5G
MOLE67209584-5GEA
0
EUR
MOLE67209584-5G
4,6-Dimethoxysalicylaldehyd
4,6-Dimethoxysalicylaldehyd
Formel:
C₉H₁₀O₄ Molecular Weight: 182,1733 g/mol |
MDL:
MFCD00003328 CAS-Nummer: 708-76-9 EINECS: 211-904-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...