3-Methylcyclopentan-1,2-dion
Lieferant: Molekula
67214708-100G
MOLE67214708-100GEA
0
EUR
MOLE67214708-100G
3-Methylcyclopentan-1,2-dion
3-Methylcyclopentan-1,2-dion
Formel:
C₆H₈O₂ Siedepunkt: 210 °C (1013 hPa) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001417 CAS-Nummer: 765-70-8 EINECS: 212-154-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...