DEAE-Dextran

Lieferant: Molekula

Synonyme: Diethylaminoethyldextran

67423750-25G
MOLE67423750-25GEA 242 EUR
MOLE67423750-25G
DEAE-Dextran
DEAE-Dextran
Lagertemperatur: Raumtemperatur
MDL: MFCD00081561
CAS-Nummer: 9015-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR