(Benzyl)triphenylphosphoniumiodid

Lieferant: Molekula

67549788-5G
MOLE67549788-5GEA 0 EUR
MOLE67549788-5G
(Benzyl)triphenylphosphoniumiodid
(Benzyl)triphenylphosphoniumiodid
Formel: C₂₅H₂₂IP
Schmelzpunkt: 250…252 °C
CAS-Nummer: 15853-35-7
EINECS: 239-977-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR