4-Brombenzylalkohol
Lieferant: Molekula
Synonyme:
(4-Bromophenyl)methanol
67559164-25G
MOLE67559164-25GEA
0
EUR
MOLE67559164-25G
4-Brombenzylalkohol
4-Brombenzylalkohol
Formel:
C₇H₇BrO Molecular Weight: 187,04 g/mol Siedepunkt: 268 °C (1013 hPa) Schmelzpunkt: 77…78 °C |
MDL:
MFCD00004650 CAS-Nummer: 873-75-6 EINECS: 212-851-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...