4,4'-Bis(dimethylamino)benzhydrol

Lieferant: Molekula

67942283-25G
MOLE67942283-25GEA 0 EUR
MOLE67942283-25G
4,4'-Bis(dimethylamino)benzhydrol
4,4'-Bis(dimethylamino)benzhydrol
Formel: C₁₇H₂₂N₂O
MDL: MFCD00008313
CAS-Nummer: 119-58-4
EINECS: 204-335-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR