TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)

Lieferant: Molekula

Synonyme: 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-Oxide Tetrafluorobo, TATU, O-(7-Azabenzotriazole-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 2-(3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate

67943083-5G
MOLE67943083-5GEA 68.7 EUR
MOLE67943083-5G
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)
Formel: C₁₀H₁₅BF₄N₆O
Molecular Weight: 322,1 g/mol
MDL: MFCD08064301
CAS-Nummer: 873798-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR