2-(4-Chlorphenoxy)isobuttersäure
Lieferant: Molekula
Synonyme:
2-(4-Chlorophenoxy)-2-methylpropionic acid, Clofibric acid, 2-(4-Chlorophenoxy)-2-methylpropanoic acid
69210117-100G
MOLE69210117-100GEA
0
EUR
MOLE69210117-100G
2-(4-Chlorphenoxy)isobuttersäure
2-(4-Chlorphenoxy)isobuttersäure
Formel:
C₁₀H₁₁ClO₃ Molecular Weight: 214,65 g/mol |
MDL:
MFCD00004192 CAS-Nummer: 882-09-7 EINECS: 212-925-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...