1,1,2-Trimethyl-1H-benz[e]indol

Lieferant: Molekula

Synonyme: 1,1,2-Trimethyl-1H-benzo[e]indole

69351493-5G
MOLE69351493-5GEA 0 EUR
MOLE69351493-5G
1,1,2-Trimethyl-1H-benz[e]indol
1,1,2-Trimethyl-1H-benz[e]indol
Formel: C₁₅H₁₅N
Molecular Weight: 209,29 g/mol
MDL: MFCD00082627
CAS-Nummer: 41532-84-7
EINECS: 255-429-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR