4-(Benzyloxy)-3-methoxybenzaldehyd

Lieferant: Molekula

70109116-5G
MOLE70109116-5GEA 0 EUR
MOLE70109116-5G
4-(Benzyloxy)-3-methoxybenzaldehyd
4-(Benzyloxy)-3-methoxybenzaldehyd
Formel: C₁₅H₁₄O₃
MDL: MFCD00003365
CAS-Nummer: 2426-87-1
EINECS: 219-379-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR