Chromotrop 2B

Lieferant: Molekula

70226119-25G
MOLE70226119-25GEA 0 EUR
MOLE70226119-25G
Chromotrop 2B
Chromotrop 2B
Formel: C₁₆H₉N₃Na₂O₁₀S₂
MDL: MFCD00003951
CAS-Nummer: 548-80-1
EINECS: 208-959-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR