(1S,2R)-(+)-2-Amino-1,2-diphenylethanol

Lieferant: Molekula

72095716-5G
MOLE72095716-5GEA 0 EUR
MOLE72095716-5G
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
Formel: C₁₄H₁₅NO
Molecular Weight: 213,28 g/mol
MDL: MFCD00074959
CAS-Nummer: 23364-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR