(Dichlormethyl)methylether

Lieferant: Molekula

Synonyme: Dichlormethyl-methylether, Dichlor(methoxy)methan

72553642-100G
MOLE72553642-100GEA 0 EUR
MOLE72553642-100G
(Dichlormethyl)methylether
(Dichlormethyl)methylether
Formel: C₂H₄Cl₂O
Molecular Weight: 114,96 g/mol
Dichte: 1,29 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00000828
CAS-Nummer: 4885-02-3
EINECS: 225-498-9
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR