2-Amino-6-methoxybenzothiazol
Lieferant: Molekula
Synonyme:
6-Methoxybenzothiazol-2-ylamine
72628801-25G
MOLE72628801-25GEA
0
EUR
MOLE72628801-25G
2-Amino-6-methoxybenzothiazol
2-Amino-6-methoxybenzothiazole
Formel:
C₈H₈N₂OS Molecular Weight: 180,23 g/mol Siedepunkt: 348 °C (1013 hPa) Schmelzpunkt: 160 °C |
MDL:
MFCD00005787 CAS-Nummer: 1747-60-0 EINECS: 217-130-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...