2,6-Diaminopurin

Lieferant: Molekula

Synonyme: 9H-Purine-2,6-diamine, 7H-Purine-2,6-diamine

73574308-5G
MOLE73574308-5GEA 0 EUR
MOLE73574308-5G
2,6-Diaminopurin
2,6-Diaminopurine
Formel: C₅H₆N₆
Molecular Weight: 150,14 g/mol
MDL: MFCD00213668
CAS-Nummer: 1904-98-9
EINECS: 217-605-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR