Methyl-2,4-dihydroxy-3,6-dimethylbenzoat

Lieferant: Molekula

Synonyme: 2,4-Dihydroxy-3,6-dimethylbenzoesäuremethylester

73626632-5G
MOLE73626632-5GEA 0 EUR
MOLE73626632-5G
Methyl-2,4-dihydroxy-3,6-dimethylbenzoat
Methyl-2,4-dihydroxy-3,6-dimethylbenzoat
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Dichte: 1 g/cm³ (20 °C)
MDL: MFCD00157202
CAS-Nummer: 4707-47-5
EINECS: 225-193-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR