4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL, freies Radikal)

Lieferant: Molekula

Synonyme: 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radical, 4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, Tempol

73749999-5G
MOLE73749999-5GEA 0 EUR
MOLE73749999-5G
4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL, freies Radikal)
4-Hydroxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Hydroxy-TEMPO, TEMPOL)
Formel: C₉H₁₈NO₂
Molecular Weight: 172.25 g/mol
MDL: MFCD00006478
CAS-Nummer: 2226-96-2
EINECS: 218-760-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR