N-Acetyl-DL-leucin
Lieferant: Molekula
Synonyme:
N-Acetyl-DL-leucine, Ac-DL-Leu-OH, 2-Acetamido-4-methylpentanoic acid
73891210-25G
MOLE73891210-25GEA
85.1
EUR
MOLE73891210-25G
N-Acetyl-DL-leucin
N-Acetyl-DL-leucin
Formel:
C₈H₁₅NO₃ Molecular Weight: 173,2 g/mol |
MDL:
MFCD00026498 CAS-Nummer: 99-15-0 EINECS: 202-734-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...