2-Chlor-5-fluoranilin

Lieferant: Molekula

74229030-5G
MOLE74229030-5GEA 0 EUR
MOLE74229030-5G
2-Chlor-5-fluoranilin
2-Chlor-5-fluoranilin
Formel: C₆H₅ClFN
MDL: MFCD00084852
CAS-Nummer: 452-83-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR