3,5-Dihydroxybenzoesäure
Lieferant: Molekula
Synonyme:
alpha-Resorcylic acid
74468622-500G
MOLE74468622-500GEA
0
EUR
MOLE74468622-500G
3,5-Dihydroxybenzoesäure
3,5-Dihydroxybenzoic acid
Formel:
C₇H₆O₄ Molecular Weight: 154,12 g/mol Schmelzpunkt: 236…238 °C Flammpunkt: >200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002512 CAS-Nummer: 99-10-5 EINECS: 202-730-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...