2-Benzoxazolinon

Lieferant: Molekula

Synonyme: benzo[d]oxazol-2(3H)-one

75203127-100G
MOLE75203127-100GEA 0 EUR
MOLE75203127-100G
2-Benzoxazolinon
2-Benzoxazolinon
Formel: C₇H₅NO₂
Molecular Weight: 135,12 g/mol
MDL: MFCD00005716
CAS-Nummer: 59-49-4
EINECS: 200-430-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR