3-(Diethylamino)phenol

Lieferant: Molekula

Synonyme: N,N-Diethyl-3-hydroxyanilin

75630880-100G
MOLE75630880-100GEA 71.6 EUR
MOLE75630880-100G
3-(Diethylamino)phenol
3-(Diethylamino)phenol
Formel: C₁₀H₁₅NO
Molecular Weight: 165,24 g/mol
Siedepunkt: 276 °C (1013 hPa)
Schmelzpunkt: 78 °C
MDL: MFCD00002265
CAS-Nummer: 91-68-9
EINECS: 202-090-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR